Please login to your online account to display your discounted pricing

EMD Millipore 13mm Nonsterile Millex™ Syringe Filters - Hydrophobic PTFE Membrane


Overmolded polypropylene housing and optimized housing design

Manufacturer: emd millipore  SLFGX13TL

 View more versions of this product

Catalog No. SLFGX13TL

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Inlet Style male luer slip
Applications For Preparation of organic solutions
Pore Size 0.2μm
Outlet Style Male Tube Outlet
Material Hydrophobic Polytetrafluoroethylene (PTFE)
Housing Material Polypropylene
Quantity 100/Pk.
Color Code Red
Type Syringe Filter FG
Diameter (Metric) 13mm