missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore 13mm Nonsterile Millex™ Syringe Filters - Durapore™ PVDF Membrane

Overmolded polypropylene housing and optimized housing design

$297.00 - $1,873.00

Specifications

Color Code Yellow
Applications For filtering aqueous and organic solutions
Diameter (Metric) 13mm
Type Syringe Filter
Housing Material HDPE
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. No. per Pack Outlet Style Pore Size Price Quantity  
Catalog Number Mfr. No. No. per Pack Outlet Style Pore Size Price Quantity  
SLGVX13NK EMD Millipore
SLGVX13NK
1000/Pk. Male Luer slip 0.22μm Pack of 1000 Each for $1,873.00
SLGVX13NL EMD Millipore
SLGVX13NL
100/Pk. Male Luer slip 0.22μm Pack of 100 Each for $297.00
SLHVX13NK EMD Millipore
SLHVX13NK
1000/Pk. Male Luer slip 0.45μm Pack of 1000 Each for $1,863.00
SLHVX13NL EMD Millipore
SLHVX13NL
100/Pk. Male Luer slip 0.45μm Pack of 100 Each for $297.00
SLGVX13TL EMD Millipore
SLGVX13TL
100/Pk. Tube outlet 0.22μm Pack of 100 Each for $308.00
SLHVX13TL EMD Millipore
SLHVX13TL
100/Pk. Tube outlet 0.45μm Pack of 100 Each for $308.00
Specifications

Description

  • Durapore™ (PVDF) membranes combine fast flow with low protein binding
  • Overmolded polypropylene housing design provides a more robust filter, with a maximum housing pressure of 150psig (10 bar) enabling solutions to be filtered faster
  • Optimized housing design maintains 50% less hold volume than traditional 13mm filters to ensure maximum sample recovery
  • Fully automated manufacturing process minimizes the risk of contamination and meets pharmaceutical standards
  • Color-coded band on housing clearly indicates membrane type
  • Order Info

    Inlet/outlet connections: female Luer-Lok™/male Luer slip.

Description & Specifications

Specifications

Yellow
13mm
HDPE
For filtering aqueous and organic solutions
Syringe Filter
Durapore (PVDF)