EMD Millipore™ 0.30μm Pore Size MF-Millipore™ Mixed Cellulose Ester Membranes
widely used in analytical/research applications.
Manufacturer:  EMD Millipore™ PHWP04700
 View more versions of this product
Catalog No. PHWP04700
Description
Description
MF-Millipore membrane is composed of mixed cellulose membrane and has a 0.3μm pore size. With biologically inert mixtures of cellulose acetate and cellulose nitrate, MF-Millipore membranes are widely used in analytical/research applications.
Specifications
Specifications
0.3μm
47mm
Hydrophilic
Mixed Cellulose Esters (MCE)
White
Documents