Please login to your online account to display your discounted pricing

EMD 66684, Tocris

Potent, selective non-peptide AT1 antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material EMD 66684
Assay Percent Range >98%
Molecular Formula C28H30N8O2·HCl
Formula Weight 547.05
Synonym 2-Butyl-3,4-dihydro-N,N-dimethyl-4-oxo-3-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl-4-yl]methyl]-5H-imidazo[4,5-c]pyridine-5-acetamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

184910

 
r & d systems
184910
10mg Each for $252.39

184950

 
r & d systems
184950
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 547.05
Quantity 10mg
Chemical Name or Material EMD 66684
Assay Percent Range >98%
Molecular Formula C28H30N8O2·HCl
CAS 1216884-39-7
Recommended Storage Desiccate at +4°C
Synonym 2-Butyl-3,4-dihydro-N,N-dimethyl-4-oxo-3-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl-4-yl]methyl]-5H-imidazo[4,5-c]pyridine-5-acetamide hydrochloride
Safety and Handling

Desiccate at +4°C