Please login to your online account to display your discounted pricing

EMD 281014 hydrochloride, Tocris™

Selective 5-HT2A antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material EMD 281014 hydrochloride
Assay Percent Range >98%
Molecular Formula C22H21FN4O·HCl
Formula Weight 412.89
Synonym 7-[[4-[2-(4-Fluorophenyl)ethyl]-1-piperazinyl]carbonyl]-1H-indole-3-carbonitrile hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

447010

 
r & d systems
447010
10mg Each for $201.36

447050

 
r & d systems
447050
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 412.89
Quantity 10mg
Chemical Name or Material EMD 281014 hydrochloride
CAS 443144-27-2
Synonym 7-[[4-[2-(4-Fluorophenyl)ethyl]-1-piperazinyl]carbonyl]-1H-indole-3-carbonitrile hydrochloride
Assay Percent Range >98%
Molecular Formula C22H21FN4O·HCl
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature