missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eluent Solution, for Ion Chromatography, 3mM NaHCO-Na2CO3, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,497-19-8,144-55-8
CAS Min % 98.0,0.02,0.02
CAS Max % 100.0,0.03,0.03
Physical Form Liquid
Chemical Name or Material Eluent Solution, 3mM NaHCO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2816-1 Ricca Chemical Company
RI28161
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
2816-32 Ricca Chemical Company
RI281632
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Ion Chromatography
Description & Specifications

Chemical Identifiers

7732-18-5,497-19-8,144-55-8
100.0,0.03,0.03
Eluent Solution, 3mM NaHCO
98.0,0.02,0.02
Liquid

Specifications

Liquid
4L
0.03,0.03,100.0
Poly natural
Clear colorless
7732-18-5,497-19-8,144-55-8
98.0,0.02,0.02
Eluent Solution, 3mM NaHCO