Please login to your online account to display your discounted pricing

Eluent Solution, for Ion Chromatography, 3mM Na 3mM Na2CO3 - 2 mM NaOH, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Eluent Solution
Name Note 3mM Na
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 497-19-8
CAS Min % 0.03
CAS Max % 0.03
CAS 1310-73-2
CAS Min % 0.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

2820-1

 
ricca chemical company
RI28201
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2820-32

 
ricca chemical company
RI282032
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Eluent Solution
CAS 497-19-8
CAS Min % 98.0
CAS Max % 0.03
CAS Min % 0.03
CAS Max % 0.02
CAS Max % 100.0
Name Note 3mM Na
CAS Min % 0.02
CAS 1310-73-2
Packaging Poly natural
CAS 7732-18-5

Ion Chromatography