Please login to your online account to display your discounted pricing

Eletriptan hydrobromide, Tocris™

Orally active, selective 5-HT1B/1D agonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material Eletriptan hydrobromide
Assay Percent Range >99%
Molecular Formula C22H26N2O2S·HBr
Formula Weight 463.43
Synonym 3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

386210

 
r & d systems
386210
10mg Each for $252.39

386250

 
r & d systems
386250
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 463.43
Quantity 10mg
CAS 177834-92-3
Assay Percent Range >99%
Molecular Formula C22H26N2O2S·HBr
Chemical Name or Material Eletriptan hydrobromide
Synonym 3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole hydrobromide
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature