missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BioTek™ BioSpa™ 8 Automated Incubators

Links BioTek washers, dispensers, readers and imagers for unattended automation of incubated workflows, including live cell assays and biochemical assays

Specifications

Description Incubator
Certifications/Compliance CE and TUV marked, RoHS compliant
Temperature Range: to 45°C, Control resolution: 0.1°C, Uniformity + 0.5°C at 37°C
Depth (Metric) 51cm
Width (Metric) 69cm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Item Description Price Quantity  
Catalog Number Mfr. No. Item Description Price Quantity  
11-120-550 Biotek Instruments Inc
BIOSPA
Incubation to 45°C, with control and monitoring N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
11-120-551 Biotek Instruments Inc
BIOSPAG
Incubation to 45°C and CO2/O2 control and monitoring, water pan and humidity monitoring N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Full unattended workflow automation integrating sample prep with detection/imaging for up to 8 plates
  • Real time temperature and CO2/O2 control and monitoring, with humidity monitoring alleviates common problems associated w/assay failure
  • Continuous recording of environment conditions and customized notifications allow detailed assay monitoring and analysis
  • Compact system integration fits in biosafety cabinet, providing the ultimate cell assay protection
  • Uncomplicated BioSpa software and simple integration for rapid implementation with short learning curve
  • Easy access for cleaning, plus HEPA filtration and lid handling station
  • Fully automated processing for up to 2 weeks for long term live cell assays
  • Handles low and high density microplates (6- to 1536-well standard height microplates, w/or w/o lids.), cell culture and Petri dishes (60 mm and 35 mm) and T25 flasks
  • Robotic arm moves plate to and from BioSpa 8 and connected instrument; handles de-lidding and re-lidding plates
  • Compatible with Cytation 5, Cytation 3, Synergy Neo2, Synergy H1, Epoch 2, EL406, 405 TS, 405 LS, MultiFlo FX, MultiFlo
Applications:
  • Automated live cell assays: wash, dispense, image
  • ELISA automation: wash, dispense, read 
  • Multiple assays: For example, run cell maintenance and live cell imaging assays in parallel
  • Automated kinetic measurements on multiple microplates in parallel

Warranty

1 year

Description & Specifications

Specifications

Incubator
Range: to 45°C, Control resolution: 0.1°C, Uniformity + 0.5°C at 37°C
69cm
CO2 Control : Range 0 – 20%, Control resolution ±0.1%, Stability ±0.2 at 5% CO2; O2 Control: Range 1 – 19%, Control resolution ±0.1%, Stability ±0.2 at 1% O2; Humidity: rH 80 to 95% (lidded plates and 5% CO2)
CE and TUV marked, RoHS compliant
51cm
48cm