missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Electrolyte Solution, 3mol/L KCl, Mettler Toledo™

$32.04 - $47.52

Chemical Identifiers

CAS 7447-40-7
Physical Form Liquid
Molecular Formula KCl
Formula Weight 74.55
Synonym Potassium Chloride
Chemical Name or Material Electrolyte Solution, 3mol/L KCl
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-917-103 Mettler Toledo
ME63056166
500mL Each for $47.52
01-917-107 Mettler Toledo
ME63056165
250mL Each for $32.04
Specifications Description & Specifications

Chemical Identifiers

7447-40-7
KCl
Potassium Chloride
Liquid
74.55
Electrolyte Solution, 3mol/L KCl

Specifications

500mL
Liquid
Potassium Chloride
Electrolyte Solution, 3mol/L KCl
74.55
7447-40-7
KCl