Please login to your online account to display your discounted pricing

Electrolyte Solution, 3mol/L KCl, AgCl Saturated, Mettler Toledo™

$30.93 - $40.63

Chemical Identifiers

Chemical Name or Material Electrolyte Solution, 3mol/L KCl, AgCl Saturated
CAS 7447-40-7
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

01-917-108

 
mettler toledo
ME63056168
250mL Each for $40.63

01-917-111

 
mettler toledo
ME63056167
125mL Each for $30.93
Description & Specifications

Specifications

Chemical Name or Material Electrolyte Solution, 3mol/L KCl, AgCl Saturated
CAS 7447-40-7
Physical Form Liquid
Quantity 250mL