missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Electrolyte Solution, 1mol/L KNO3, Mettler Toledo™

$36.93 - $54.00

Chemical Identifiers

CAS 7757-79-1
Physical Form Liquid
Molecular Formula KNO3
Formula Weight 101.10
Synonym Potassium Chloride
Chemical Name or Material Electrolyte Solution, 1mol/L KNO3
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-917-105 Mettler Toledo
ME63056172
500mL Each for $54.00
01-917-109 Mettler Toledo
ME63056171
250mL Each for $53.14
01-917-112 Mettler Toledo
ME63056170
125mL Each for $36.93
Specifications Description & Specifications

Chemical Identifiers

7757-79-1
KNO3
Potassium Chloride
Liquid
101.10
Electrolyte Solution, 1mol/L KNO3

Specifications

7757-79-1
KNO3
Potassium Chloride
Electrolyte Solution, 1mol/L KNO3
Liquid
101.10
500mL