Please login to your online account to display your discounted pricing

Electrolyte Solution, 1mol/L KNO3, Mettler Toledo™


Quantity 125mL

Manufacturer: mettler toledo  ME63056170

 View more versions of this product

Catalog No. 01-917-112

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 125mL
Formula Weight 101.10
Physical Form Liquid
Chemical Name or Material Electrolyte Solution, 1mol/L KNO3
CAS 7757-79-1
Molecular Formula KNO3
Synonym Potassium Chloride

Specifications

Chemical Name or Material Electrolyte Solution, 1mol/L KNO3
CAS 7757-79-1
Physical Form Liquid
Molecular Formula KNO3
Formula Weight 101.10
Synonym Potassium Chloride