missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Electrolyte Solution, 1M LiCl in Ethanol, Mettler Toledo™

$83.12 - $115.26

Chemical Identifiers

Physical Form Liquid
Molecular Formula LiCl
Synonym Lithium Chloride
Chemical Name or Material Electrolyte Solution, 1M LiCl in Ethanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-917-113 Mettler Toledo
ME63056173
125mL Each for $83.12
01-917-110 Mettler Toledo
ME63056174
250mL Each for $115.26
Specifications Description & Specifications

Chemical Identifiers

Liquid
LiCl
Lithium Chloride
Electrolyte Solution, 1M LiCl in Ethanol

Specifications

Liquid
Lithium Chloride
Electrolyte Solution, 1M LiCl in Ethanol
LiCl
125mL