Please login to your online account to display your discounted pricing

Electrode Fill Solution, For Nitrate, LabChem™

$66.69 - $66.69

Chemical Identifiers

Chemical Name or Material Electrode Fill Solution
CAS 7732-18-5
CAS Max % 99.47
CAS 7783-20-2
CAS Max % 0.53
Name Note For Nitrate
Linear Formula (NH4)2SO4
Formula Weight 132.13
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC178901

 
lab chem
LC178901
500mL Each for $66.69
Description & Specifications

Specifications

Formula Weight 132.13
Chemical Name or Material Electrode Fill Solution
Viscosity 1.01cSt
Decomposition Information Gaseous Ammonia; Sulfur compounds
Color Clear, colorless
Quantity 500mL
Name Note For Nitrate
Physical Form Liquid
Linear Formula (NH4)2SO4
Density 1g/mL
CAS 7732-18-5
CAS Max % 99.47
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 7783-20-2
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 0.53
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.