missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Elcatonin

For cellular and molecular biology applications

Specifications

Product Type Elcatonin
Molecular Weight 3349.74
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66874EXD View Documents Alfa Aesar™
J66874EXD
0.5mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66874MCR View Documents Alfa Aesar™
J66874MCR
1mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

Elcatonin
-30° to -10°C
RUO
3349.74
Solid
HPLC

For Research Use Only