missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Elastase Substrate IV, Colorimetric

For cellular and molecular biology applications

Specifications

Substrate Type Colorimetric
Storage Requirements -30° to -10°C
Color White to off-white
Molecular Weight 562.60
Format Lyophilized
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
AAJ66363EXD Alfa Aesar™
J66363EXD
0.5mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66363MCR Alfa Aesar™
J66363MCR
1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Specifications

Colorimetric
White to off-white
Lyophilized
-30° to -10°C
562.60