Please login to your online account to display your discounted pricing

Elastase Inhibitor II

A potent irreversible inhibitor of human neutrophil elastase

$162.73 - $162.73

Specifications

Formula Weight 474.90g/mol
Synonym HNE Inhibitor; MeOSuc-AAPA-CMK
Storage Requirements −30° to −10°C
Format Solid
Sensitivity Moisture Sensitive
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65641LB0

 
alfa aesar™
J65641LB0
Each for $162.73
Description & Specifications

Specifications

Formula Weight 474.90g/mol
Synonym HNE Inhibitor; MeOSuc-AAPA-CMK
Storage Requirements −30° to −10°C
Format Solid
Sensitivity Moisture Sensitive
For Use With (Application) A potent irreversible inhibitor of human neutrophil elastase
Description White
Solubility Soluble in methanol or DMSO