missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Elaidic Anhydride, TCI America™

Manufacturer:  TCI America E04181G

 View more versions of this product

Catalog No. E04181G



Specifications

Specifications

Elaidic Anhydride
46°C
55726-25-5
MFCD00070026
OCNZHGHKKQOQCZ-XPWSMXQVSA-N
[(E)-octadec-9-enoyl] (E)-octadec-9-enoate
6183559
Crystal-Powder at 20°C
White-Yellow
1g
C36H66O3
trans-9-Octadecenoic Anhydride
CCCCCCCCC=CCCCCCCCC(=O)OC(=O)CCCCCCCC=CCCCCCCCC
546.921
546.92
≥98.0% (T)

Chemical Identifiers

55726-25-5
546.921
OCNZHGHKKQOQCZ-XPWSMXQVSA-N
6183559
CCCCCCCCC=CCCCCCCCC(=O)OC(=O)CCCCCCCC=CCCCCCCCC
C36H66O3
MFCD00070026
trans-9-Octadecenoic Anhydride
[(E)-octadec-9-enoyl] (E)-octadec-9-enoate
Safety and Handling

Safety and Handling

TSCA : No

Documents