Please login to your online account to display your discounted pricing

EIT hydrobromide, Tocris


Selective iNOS inhibitor, acts arginine binding site

Quantity 50mg

Manufacturer: r & d systems  RDS087350

 View more versions of this product

Catalog No. 087350

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 185.08
Recommended Storage Store at Room Temperature
Molecular Formula C3H8N2S·HBr
Chemical Name or Material EIT hydrobromide
CAS 1071-37-0
Synonym (S)-Ethylisothiourea hydrobromide
Assay Percent Range >98%

Specifications

Chemical Name or Material EIT hydrobromide
Assay Percent Range >98%
Molecular Formula C3H8N2S·HBr
Formula Weight 185.08
Synonym (S)-Ethylisothiourea hydrobromide
Accessories

Store at Room Temperature