Please login to your online account to display your discounted pricing

EIT hydrobromide, Tocris

Selective iNOS inhibitor, acts arginine binding site

$121.23 - $509.13

Chemical Identifiers

Chemical Name or Material EIT hydrobromide
Assay Percent Range >98%
Molecular Formula C3H8N2S·HBr
Formula Weight 185.08
Synonym (S)-Ethylisothiourea hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

087310

 
r & d systems
RDS087310
10mg Each for $121.23

087350

 
r & d systems
RDS087350
50mg Each for $509.13
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 185.08
Chemical Name or Material EIT hydrobromide
CAS 1071-37-0
Synonym (S)-Ethylisothiourea hydrobromide
Recommended Storage Store at Room Temperature
Molecular Formula C3H8N2S·HBr
Assay Percent Range >98%
Safety and Handling

Store at Room Temperature