missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Wood Dynamics Carts
Ideal for demonstrating Newton's laws of motion.
$239.65 - $239.65
Specifications
Product Type | Dynamics Trolley |
---|---|
Material | Wood |
For Use With (Application) | Demonstrate Newton's laws of motion |
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog Number | Mfr. No. | Weight (English) | Weight (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Weight (English) | Weight (Metric) | Price | Quantity | ||||
S96176
|
Eisco™
PH0350B |
5.7 lb. | 2.6 kg |
Each for $239.65
|
|
||||
Description
- Carts made with low-friction ball-bearing wheels
- Top surface can be loaded with additional masses
- One cart in each set is equipped with an “exploder piston,” activated by a trigger on top that can be preset to three positions
- Spring-steel hoop bumpers used for collision and energy experiments
Order Info
Sold as a set of two carts.
Specifications
Dynamics Trolley | |
Wood | |
Demonstrate Newton's laws of motion |