EISCO Student Basic Organic Chemistry Molecular Model Sets
Let students visualize three-dimensional molecule structures and understand molecular formulas.
$42.55 - $59.10
Specifications
For Use With (Application) Basic organic chemistry
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
S96538 EISCO
SET00602
54 Pieces Molecular Model Set Each for $42.55
S96539 EISCO
SET00603
114 Pieces Molecular Model Set Each for $59.10
Specifications
Description
  • Hard plastic balls of different colors represent atoms
  • Each has holes at correct angles to be connected with flexible plastic “bonds”
  • Medium links represent single covalent bonds while long links can be used for double and triple covalent bonds
54-Piece Set
  • 14 White hydrogen (1 hole)
  • Six black carbon (4 holes)
  • Two blue nitrogen (4 holes)
  • Six red oxygen (2 holes)
  • Two green halogen (1 hole)
  • 28mm Gray links (20)
  • 43mm Gray links (4)
114-Piece Set
  • 12 Black carbon (4 holes)
  • 20 White hydrogen (1 hole)
  • Six red oxygen (2 holes)
  • Four blue nitrogen (4 holes)
  • Two yellow sulphur (6 holes; 1 hole)
  • One purple phosphorus (4 holes)
  • One gray metal (1 hole)
  • 28mm Gray links (26)
  • 43mm Gray links (12)
  • 28mm White links (26)
  • Assembly tool
Description & Specifications
Specifications
Basic organic chemistry