missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Spectrum Tube Power Supply

The main use of this safe, economical and fun spectrum tube power supply is to provide a view of gas discharge spectral lines that can be viewed with something as simple as a diffraction grating, to as complicated as digital spectrometers.

Manufacturer:  Eisco™ PH1197A

Catalog No. S96205



Description

Description

  • Black panel behind the tube eliminates distracting ambient light and protects tube from breakage
  • Specially designed box holds tube firmly, preventing student from touching electrodes
  • Suitable for 10.25 in. (26cm) long spectrum tubes

Order Info

Operates on 120VAC 60Hz ±10%.

Specifications

Specifications

Power Supply
120V
CSA, CE, CUS
60Hz
1A
120V 60Hz
Documents