EISCO Ripple Tank
Allows all types of waves and wave motion.
$487.40 - $746.40
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S99348 EISCO
PH0767C
Each for $746.40
S99347 EISCO
PH0767A
Each for $487.40
Specifications
Description
  • With illuminator lamp
  • The unique design and size allows it to be used on an overhead projector
  • Complete with accessories
S99347 Ripple tank, 15.8 × 15.8 in. (40 × 40cm); push fit legs; rippler motor with eccentric cam, mounted on PVC bar; three supports for illuminator and the motor unit; motor hanging clamps; springs for motor support; halogen lamp unit, 6V; hand stroboscope; concave perspex plate; convex perspex plate; rectangular perspex plate; curved reflector; large barrier; small barrier, plastic roller bar
S99348
Controller only