Please login to your online account to display your discounted pricing
EISCO Polypropylene Cylinders
Base provides good stability.
Manufacturer: eisco  FSC1044
 View more versions of this product
Catalog No. S95179A
  • / Pack of 8 Each
Description & Specifications
Specifications
Autoclavable Yes
Material PP (Polypropylene)
Capacity (Metric) One each of: 10, 25, 50, 100, 250, 500, 1000, 2000mL
Includes 10, 25, 50, 100, 250, 500, 1000, 2000mL cylinders
Quantity 8/Set