missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Polypropylene Beakers with Spout

These polypropylene beakers have excellent optical clarity, very good chemical resistance, and are autoclavable. There are raised graduations and they have tapered spouts.

Manufacturer:  Eisco™ CH0137BF

 View more versions of this product

Catalog No. S97808B



Description

Description

  • Autoclavable
  • Raised graduations on outer surface
  • Tapered spout
Specifications

Specifications

100 mL
For Excellent Clarity and Very Good Chemical Resistance
9.98 cm
9.44 in.
12
Beaker
18.99 cm
Yes
Euro
Polypropylene
3.93 in.
Spout
24 cm
12/Pk.
7.48 in.
Autoclavable
Raised Outer Graduated Scale
SDS
Documents
Product Certifications
Promotions

Promotions