missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Polypropylene Beakers with Spout

These polypropylene beakers have excellent optical clarity, very good chemical resistance, and are autoclavable. There are raised graduations and they have tapered spouts.

Manufacturer:  Eisco™ CH0137AF

 View more versions of this product

Catalog No. S97808A



Description

Description

  • Autoclavable
  • Raised graduations on outer surface
  • Tapered spout
Specifications

Specifications

1.69 oz.
Polypropylene
7.48 in.
Spout
22 cm
12/Pk.
7.08 in.
Autoclavable
Raised Outer Graduated Scale
50 mL
For Excellent Clarity and Very Good Chemical Resistance
18.99 cm
8.66 in.
12
Beaker
17.98 cm
Yes
Euro
SDS
Documents
Product Certifications
Promotions

Promotions