EISCO Polypropylene Beakers with Spout
Excellent clarity and very good chemical resistance.
$15.45 - $95.75
Specifications
Autoclavable Yes
Material PP (Polypropylene)
Product Type Beaker
For Use With (Application) For excellent clarity and very good chemical resistance
Graduated Yes
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S97808A EISCO
CH0137AF
50mL Dozen of 12 Pair for $20.15
S97808B EISCO
CH0137BF
100mL Dozen of 12 Pair for $15.45
S97808C EISCO
CH0137DF
500mL Dozen of 12 Pair for $40.60
S97808E EISCO
CH0137FF
2000mL Dozen of 12 Pair for $95.75
S97808D EISCO
CH0137EF
1000mL Dozen of 12 Pair for $53.00
S97808F EISCO
CH037GSET
One each: 50, 100, 250, 500, 1000, 2000mL Pack of 6 Each for $32.00
Specifications
Description
  • Autoclavable
  • Raised graduations on outer surface
  • Tapered spout
Description & Specifications
Specifications
Yes
Beaker
Yes
Raised outer graduated
PP (Polypropylene)
For excellent clarity and very good chemical resistance
Spout
Euro