Eisco™ Optical Lens
Eisco lenses are spherical, optically worked glass with ground edges and highly polished making them ideal for classroom experiments.
Manufacturer: Eisco™ FSCI029
Description
- Optical lens
- Choice of double convex or concave
- Choice of diameter and focal length
Specifications
Each | |
3 in. | |
20cm | |
Ground edges, highly polished |
7.5cm | |
7.8 in. | |
Spherical | |
Double concave lens |