Please login to your online account to display your discounted pricing
Lab-Aids™ Energy Transfer: Waves, Sound and Light Kit
Students use water and springs to produce both longitudinal and transverse mechanical waves, then compare their similarities and differences.
Manufacturer: lab-aids™  211EL
 View more versions of this product
Catalog No. S98797A
  • / Each
Description & Specifications
Specifications
Includes Mini wave tank, one spring, one elastic string with barbed ends, one plastic paddle, one 15cm ruler
For Use With (Application) Studying wavelengths and frequencies
Product Type Extender Components Kit