missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Goggle Sanitizer Cabinet
UV light kills the germs on goggles after laboratory use.
Manufacturer: Eisco™ GGSN10
Description
- Wall-mounted or bench top unit runs on 120VAC
- Heavy gauge CRC steel treated with white enamel finish
- Unit has doors and includes 15 minute timer with UV bulb
- UV light operates only when doors are closed
- Light leak avoided by tight fitting piano style hinges
- Sterilization process is indicated by indicator light
- Use to sanitize 35 pairs
Specifications
66.04 x 25.4 x 73.66 cm (26 x 10 x 29 in.) | |
Sanitizes up to 35 goggles at a time for efficiency | |
110V | |
21.77kg |
Equipment sterilization | |
Steel | |
48 lbs. |