missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Ceramic-Centered Iron Wire Gauze
Eisco Iron Wire Gauze with Ceramic Center is high quality and extra strong for long-term use in the classroom laboratory.
Manufacturer: Eisco™ CH0525B
Description
- Squares of extra-strong quality
- Pack of 10
Specifications
5.9 in. | |
6 x 6 x 1.1 in. (15.24 x 15.24 x 3cm) | |
3 cm | |
6 in. | |
Square | |
15.24 cm | |
Wire Gauze |
15 cm | |
1.1 in. | |
Ceramic center | |
10 Pack | |
6 in. | |
Iron | |
15.24 cm |