missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Ceramic-Centered Iron Wire Gauze
Eisco Iron Wire Gauze with Ceramic Center is high quality and extra strong for long-term use in the classroom laboratory.
Manufacturer: Eisco™ CH0525A
Description
- Squares of extra-strong quality
- Pack of 10
Specifications
12.7 cm | |
Wire Gauze | |
Ceramic Center | |
12.5 cm | |
Folded | |
11 cm | |
5 in. | |
Square |
Ceramic, Iron | |
12.7 cm | |
4.9 in. | |
5 x 5 x 4.3 in. (12.7 x 12.7 x 11cm) | |
4.3 in. | |
Ceramic center | |
10 Pack | |
5 in. |