Please login to your online account to display your discounted pricing

EIPA, Tocris

Inhibits TRPP3-mediated currents; also inhibits the Na+/H+ exchanger (NHE)

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material EIPA
Assay Percent Range >98%
Molecular Formula C11H8ClN7O
Formula Weight 299.76
Synonym 3-Amino-N-(aminoiminomethyl)-6-chloro-5-[ethyl(1-methylethyl)amino]-2-pyrazinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

337810

 
r & d systems
337810
10mg Each for $113.57

337850

 
r & d systems
337850
50mg Each for $478.51
Description & Specifications

Specifications

Formula Weight 299.76
Quantity 10mg
Chemical Name or Material EIPA
Assay Percent Range >98%
Molecular Formula C11H8ClN7O
CAS 1154-25-2
Synonym 3-Amino-N-(aminoiminomethyl)-6-chloro-5-[ethyl(1-methylethyl)amino]-2-pyrazinecarboxamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C