Invitrogen™ EIF2AK2 (PKR) Recombinant Human Protein DFS Item
EIF2AK2 (PKR) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha.
Manufacturer:  Invitrogen™ LSPV4822
 View more versions of this product
Catalog No. PV4822
Description
Description
EIF2AK2 (PKR) phosphorylates Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses. EIF2AK2 plays a key role in antiviral defense mediated through its direct activation by double-stranded RNA produced by viral infection.
Specifications
Specifications
NP_002750.1
Frozen
62.2kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
PEK Kinase Family
Human
GST Tag
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
50mM Tris, pH 7.5, 50% glycerol, 0.5mM EDTA, 0.02% Triton X-100, 150mM NaCl, 2mM DTT
1mg
RUO - research use only
EIF2AK2 (PKR)
Baculovirus
aa 252-551
Yes
Documents