missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ EIF2AK1 (HRI) Human Protein
EIF2AK1 (HRI) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha
Manufacturer: Invitrogen™ LSPV5857
Description
EIF2AK1 (HRI) specifically phosphorylates Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK1, signaling is initiated by low levels of heme.
Specifications
NP_055228.2 | |
Frozen | |
1mg | |
RUO - research use only | |
EIF2AK1 (HRI) | |
Baculovirus | |
Human |
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis | |
98.6kDa | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Full Length | |
Kinase Assay | |
PEK Kinase Family | |
GST Tag |