missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eicosane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 204371000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Eicosane | |
36.0°C to 38.0°C | |
0.7886g/mL | |
>112°C | |
98.5% min. (GC) | |
C20H42 | |
MFCD00009344 | |
01, 174 | |
eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane | |
CBFCDTFDPHXCNY-UHFFFAOYSA-N | |
icosane | |
8222 | |
282.54 | |
Crystalline Solid |
112-95-8 | |
White | |
220.0°C (30.0 mmHg) | |
Authentic | |
Glass bottle | |
CH3(CH2)18CH3 | |
100 g | |
0.7886 | |
Solubility in water: insoluble. Other solubilities: soluble in ether | |
CCCCCCCCCCCCCCCCCCCC | |
282.56 | |
CHEBI:43619 | |
99% |
Chemical Identifiers
112-95-8 | |
282.56 | |
CBFCDTFDPHXCNY-UHFFFAOYSA-N | |
8222 | |
icosane |
C20H42 | |
MFCD00009344 | |
eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane | |
CHEBI:43619 | |
CCCCCCCCCCCCCCCCCCCC |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 204-018-1
RUO – Research Use Only