missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eicosane, 99%, ACROS Organics™

Manufacturer:  Acros Organics 204370020

 View more versions of this product

Catalog No. AC204370020



Specifications

Specifications

Eicosane
>112°C
Glass bottle
220.0°C (30.0 mmHg)
36.0°C to 38.0°C
112-95-8
C20H42
MFCD00009344
eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane
CBFCDTFDPHXCNY-UHFFFAOYSA-N
icosane
8222
282.54
99%
0.7886g/mL
Authentic
0.7886
White
2g
99%
CH3(CH2)18CH3
01, 174
Solubility in water: insoluble. Other solubilities: soluble in ether
CCCCCCCCCCCCCCCCCCCC
282.556
CHEBI:43619
Crystalline Solid

Chemical Identifiers

112-95-8
282.556
CBFCDTFDPHXCNY-UHFFFAOYSA-N
8222
icosane
C20H42
MFCD00009344
eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane
CHEBI:43619
CCCCCCCCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

GHS P Statement May be fatal if swallowed and enters airways.

GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting.

SDS Documents