Please login to your online account to display your discounted pricing

EHT 1864, Tocris

Potent inhibitor of Rac family GTPases

$266.69 - $1,121.62

Chemical Identifiers

Physical Form Solid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

387250

 
r & d systems
RDS387250
50mg Each for $1,121.62

387210R

 
r & d systems
RDS387210
10mg Each for $266.69
Description & Specifications

Specifications

Formula Weight 581.47
Quantity 50mg
Physical Form Solid
Molecular Formula C25H27F3N2O4S·2HCl
Assay Percent Range >98%
CAS 754240-09-0
Recommended Storage Store at +4°C
Synonym 5-(5-(7-(Trifluoromethyl)quinolin-4-ylthio)pentyloxy)-2-(morpholinomethyl)-4H-pyran-4-one dihydrochloride
Chemical Name or Material EHT 1864
Safety and Handling

Store at +4°C