missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ehrlich's Reagent, LabChem™

$142.57 - $142.57

Chemical Identifiers

CAS 71-23-8,100-10-7
CAS Max % 95,5
Linear Formula C9H11NO
Formula Weight 149.19
UN Number UN1993
Chemical Name or Material Ehrlich's Reagent
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC140802 Lab Chem
LC140802
1L Each for $142.57
Specifications Description & Specifications

Chemical Identifiers

71-23-8,100-10-7
C9H11NO
UN1993
95,5
149.19
Ehrlich's Reagent

Specifications

71-23-8,100-10-7
Liquid
149.19
Passes Test
Soluble in water
Carbon monoxide; Carbon dioxide; Nitrogen oxides
1L
95,5
C9H11NO
UN1993
Amber Glass
Room Temperature
Clear, yellow
Ehrlich's Reagent

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a cool, well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

Danger