missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ehrlich Aldehyde Reagent, Alcoholic, for Indole Test, Ricca Chemical

Chemical Identifiers

CAS 64-17-5,7647-01-0,7732-18-5,100-10-7,7732-18-5 (Water, Deionized)
CAS Min % 70.0,22.96,3.89,0.89
CAS Max % 73.0,23.89,4.05,0.93
Physical Form Liquid
Chemical Name or Material Ehrlich Aldehyde Reagent
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2790-16 SDS Ricca Chemical Company
RI279016
Glass amber 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
2790-4 Ricca Chemical Company
RI27904
Glass amber 120mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

For Indole Test
Description & Specifications

Chemical Identifiers

64-17-5,7647-01-0,7732-18-5,100-10-7,7732-18-5 (Water, Deionized)
73.0,23.89,4.05,0.93
Ehrlich Aldehyde Reagent
70.0,22.96,3.89,0.89
Liquid

Specifications

64-17-5,7647-01-0,7732-18-5,100-10-7,7732-18-5 (Water, Deionized)
73.0,23.89,4.05,0.93
Glass amber
Clear pale yellow
Ehrlich Aldehyde Reagent
70.0,22.96,3.89,0.89
Liquid
Alcoholic Solution
500mL