Please login to your online account to display your discounted pricing

EHNA hydrochloride, Tocris


Adenosine deaminase inhibitor

Quantity 10mg

Manufacturer: r & d systems  126110

 View more versions of this product

Catalog No. 126110

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 313.83
Quantity 10mg
CAS 81408-49-3
Molecular Formula C14H23N5O·HCl
Recommended Storage Store at Room Temperature
Synonym erythro-9-(2-Hydroxy-3-nonyl)adenine hydrochloride
Chemical Name or Material EHNA hydrochloride
Assay Percent Range >98%

Specifications

Chemical Name or Material EHNA hydrochloride
Assay Percent Range >98%
Molecular Formula C14H23N5O·HCl
Formula Weight 313.83
Synonym erythro-9-(2-Hydroxy-3-nonyl)adenine hydrochloride
Accessories

Store at Room Temperature