Please login to your online account to display your discounted pricing

EgR1 Rat anti-Human, PE, Clone: HEGR1DS, eBioscience™

Rat Monoclonal Antibody

$201.55 - $477.05

Specifications

Antigen EgR1
Clone HEGR1DS
Host Species Rat
Gene Alias Zif268, NGFI-A, KROX-24, ZENK, Znf255
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122469

 
life technologies
LS12985141
25 tests Each for $201.55

501122470

 
life technologies
LS12985142
100 tests Each for $477.05
Description & Specifications

Specifications

Antigen EgR1
Clone HEGR1DS
Host Species Rat
Gene Alias Zif268, NGFI-A, KROX-24, ZENK, Znf255
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

The monoclonal antibody HEGR1DS recognizes human Egr1 (Early Growth Response 1), a C2H2-type zinc-finger transcription factor broadly expressed in various types of tissues. Egr1 is induced within minutes after cell activation and it regulates expression of numerous factors involved in cell division, growth, and differentiation. It has been shown to suppress tumorigenesis by inducing apoptosis of cancer cells but it can also promote their proliferation. Egr1 is often used as an early cell activation marker of T and B lymphocytes as well as myeloid cells.