Please login to your online account to display your discounted pricing

EGLU, Tocris

Highly selective group II mGlu antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material EGLU
Assay Percent Range >95%
Molecular Formula C7H13NO4
Formula Weight 175.18
Synonym (2S)-α-Ethylglutamic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

097110

 
r & d systems
RDS097110
10mg Each for $201.36

097150

 
r & d systems
RDS097150
50mg Each for $844.73
Description & Specifications

Specifications

Chemical Name or Material EGLU
CAS 170984-72-2
Recommended Storage Store at -20°C
Assay Percent Range >95%
Molecular Formula C7H13NO4
Formula Weight 175.18
Synonym (2S)-α-Ethylglutamic acid
Quantity 10mg
Safety and Handling

Store at -20°C