EGFR (ErbB1) [L858R] Recombinant Human Protein, Invitrogen™ DFS Item
The epidermal growth factor receptor is a receptor tyrosine kinases that signals in response to various growth factors.
Manufacturer:  Life Technologies LSPV4289
 View more versions of this product
Catalog No. PV4289
Description
Description
Overexpression of EGFR has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. The EGFR L858R mutation is implicated in epithelial tumor formation.
Specifications
Specifications
NP_005219.2; L858R
Kinase Assay
Frozen
90.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
EGF Receptor (ErbB) Kinase Family
Yes
Baculovirus
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
EGFR (ErbB1) L858R
Human
GST Tag