missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EGFR (ErbB1) [L858R] Recombinant Human Protein, Invitrogen™ DFS Item

The epidermal growth factor receptor is a receptor tyrosine kinases that signals in response to various growth factors.

$2,035.00 - $8,670.00

Specifications

Format Frozen
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR7447A
View Documents
Life Technologies
LSPR7447A
100μg Each for $2,035.00
 
PV4289
View Documents
Life Technologies
LSPV4289
1mg Each for $8,670.00
 
Description

Description

Overexpression of EGFR has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. The EGFR L858R mutation is implicated in epithelial tumor formation.

Specifications

Specifications

Frozen
Human
GST Tag
Yes