Please login to your online account to display your discounted pricing

EGFL7 Recombinant Protein, Abnova

Human EGFL7 full-length ORF recombinant protein with GST-tag at N-terminal

$622.91 - $622.91

Specifications

Cross Reactivity Human
Format Solution
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
Molecular Weight 53.24
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

89-012-814

 
abnova corporation
H00051162P0125
25μg Each for $622.91
Description & Specifications

Specifications

Cross Reactivity Human
Format Solution
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
Molecular Weight 53.24
Name Human EGFL7 Full-length ORF Recombinant Protein with GST-tag at N-terminal
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE stained with Coomassie Blue
Recombinant Yes
Source Wheat Germ (in vitro)
Storage Requirements -80°C

  • Theoretical MW (kDa): 53.24
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array