missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
EFEMP1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002202B01P
Description
Sequence: MLKALFLTMLTLALVKSQDTEETITYTQCTDGYEWDPVRQQCKDIDECDIVPDACKGGMKCVNHYGGYLCLPKTAQIIVNNEQPQQETQPAEGTSGATTGVVAASSMATSGVLPGGGFVASAAAVAGPEMQTGRNNFVIRRNPADPQRIPSNPSHRIQCAAGYEQSEHNVCQDIDECTAGTHNCRADQVCINLRGSFACQCPPGYQKRGEQCVDIDECTIPPYCHQRCVNTPGSFYCQCSPGFQLAANNYTCVDINECDASNQCAQQCYNILGSFICQCNQGYELSSDRLNCEDIDECRTSSYLCQYQCVNEPGKFSCMCPQGYQVVRSRTCQDINECETTNECREDEMCWNYHGGFRCYPRNPCQDPYILTPENRCVCPVSNAMCRELPQSIVYKYMSIRSDRSVPSDIFQIQATTIYANTINTFRIKSGNENGEFYLRQTSPVSAMLVLVKSLSGPREHIVDLEMLTVSSIGTFRTSSVLRLTIIVGPFSFSpecifications
EFEMP1 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
NM_004105.3 | |
EFEMP1 | |
EFEMP1 (NP_004096.2, 1 a.a. ∼ 493 a.a) full-length human protein. | |
RUO | |
2202 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
EGF-containing fibulin-like extracellular matrix protein 1 | |
DHRD/DRAD/FBLN3/FBNL/FLJ35535/MGC111353/MLVT/MTLV/S1-5 | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |