Invitrogen™ EEF2K Recombinant Human Protein DFS Item
EEF2K is a highly conserved serine/threoning kinase in the calmodulin signaling pathway.
Manufacturer:  Invitrogen™ LSPV4560
 View more versions of this product
Catalog No. PV4560
Description
Description
EEF2K activity is increased in cancers, including breast cancer.
Specifications
Specifications
NP_037434
Frozen
29904
1mg
RUO - research use only
EEF2K, eEF-2K
E. coli
Human
GST Tag
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
109.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
Alpha Kinase Family
Yes
Documents